KEY151861594 Key Organics / BIONET

tert-butyl pyrazolidine-1-carboxylate

  • MDL number MFCD09817426
  • Cas Number : 57699-91-9
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94923

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BB-0207_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart