KEY151861596 Key Organics / BIONET

2-(chloromethyl)-1-methyl-1H-imidazole

  • MDL number MFCD00234119
  • Cas Number : 19225-92-4
  • Molecular Weight : 130.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182 - 184
Purity 95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100872

Safety Information

signalword Warning
pcodes P232,P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KB-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart