KEY151861597 Key Organics / BIONET

1-[(3,4-dichlorophenyl)methyl]piperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD03425739
  • Cas Number : 451485-54-4
  • Molecular Weight : 324.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260 - 262
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69401

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart