KEY151861624 Key Organics / BIONET

2-(4-bromophenyl)butanoic acid

  • MDL number MFCD01737245
  • Cas Number : 99070-18-5
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 83
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108156

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart