KEY151861634 Key Organics / BIONET

methyl N-(2,2,4-trimethyl-3-oxo-1-phenylpentyl)carbamate

  • MDL number MFCD09864844
  • Cas Number : 1000018-26-7
  • Molecular Weight : 277.364

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108181

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2640_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart