KEY151861637 Key Organics / BIONET

1-(2-nitrobenzenesulfonyl)piperazine

  • MDL number MFCD00227564
  • Cas Number : 301331-16-8
  • Molecular Weight : 271.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96816

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FB-0723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart