KEY154404370 Key Organics / BIONET

1-{1-[(4-bromophenyl)methyl]-5-methyl-1H-1,2,3-triazol-4-yl}ethan-1-ol

  • MDL number MFCD09865012
  • Cas Number : 1017782-60-3
  • Molecular Weight : 296.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97598

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/H-20195_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart