KEY154404374 Key Organics / BIONET

(3-amino-4-fluorophenyl)methanol

  • MDL number MFCD09865016
  • Cas Number : 227609-86-1
  • Molecular Weight : 141.145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 78
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101726

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LB-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart