KEY154404376 Key Organics / BIONET

1-methanesulfonylpiperidine-4-carbonyl chloride

  • MDL number MFCD09865018
  • Cas Number : 241138-40-9
  • Molecular Weight : 225.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 83
Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101728

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LB-0726_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart