KEY154404390 Key Organics / BIONET

2-[3-(2-aminophenyl)-3-oxopropyl]-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD09865034
  • Cas Number : 17515-32-1
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108189

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2959_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart