KEY154404445 Key Organics / BIONET

(2Z)-3-(dimethylamino)-2-(thiophen-3-yl)prop-2-enenitrile

  • MDL number MFCD00138240
  • Cas Number : 165404-39-7
  • Molecular Weight : 178.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 76
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10B-113_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart