KEY154404458 Key Organics / BIONET

3-methyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD01114751
  • Cas Number : 17467-35-5
  • Molecular Weight : 115.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97635

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HA-0217_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart