KEY154404491 Key Organics / BIONET

2-(4-methoxyphenoxy)-1-ethanol

  • MDL number MFCD00016568
  • Cas Number : 5394-57-0
  • Molecular Weight : 168.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108159

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2122_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart