KEY154404509 Key Organics / BIONET

2-(5-fluoro-2-methyl-1H-inden-3-yl)acetic acid

  • MDL number MFCD01076194
  • Cas Number : 32004-66-3
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-fluoro-2-methyl-1H-inden-3-yl)acetic acid
Inchi Code 1S/C12H11FO2/c1-7-4-8-2-3-9(13)5-11(8)10(7)6-12(14)15/h2-3,5H,4,6H2,1H3,(H,14,15)
Inchi Key QDDPPRDVFIJASZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108194

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-2964_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart