KEY158583295 Key Organics / BIONET

3-[5-(trifluoromethyl)pyridin-2-yl]benzoic acid

  • MDL number MFCD09972206
  • Cas Number : 946409-32-1
  • Molecular Weight : 267.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Z-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart