KEY158583296 Key Organics / BIONET

4-[(3,4-difluorophenyl)methoxy]benzaldehyde

  • MDL number MFCD09972208
  • Cas Number : 1017782-47-6
  • Molecular Weight : 248.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 88 - 90
Iupac Name 4-[(3,4-difluorobenzyl)oxy]benzaldehyde
Inchi Code 1S/C14H10F2O2/c15-13-6-3-11(7-14(13)16)9-18-12-4-1-10(8-17)2-5-12/h1-8H,9H2
Inchi Key ZTEWSBQTOYYFFO-UHFFFAOYSA-N

Description

Melting Point 88 - 90
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94484

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AC-0700_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart