KEY158583299 Key Organics / BIONET

ethyl 1-(4-amino-3-methoxyphenyl)piperidine-4-carboxylate

  • MDL number MFCD09972211
  • Cas Number : 1017782-48-7
  • Molecular Weight : 278.352

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 77
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94488

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart