KEY158583301 Key Organics / BIONET

{4-[(3,4-dichlorophenoxy)methyl]phenyl}methanol

  • MDL number MFCD09972212
  • Cas Number : 1017782-50-1
  • Molecular Weight : 283.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 97
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart