KEY158583303 Key Organics / BIONET

ethyl 2-(2,4-difluoro-5-nitrophenoxy)acetate

  • MDL number MFCD09972214
  • Cas Number : 179011-36-0
  • Molecular Weight : 261.181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47 - 49
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0718_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart