KEY158583304 Key Organics / BIONET

[(5-chloro-1,3-benzoxazol-2-yl)methyl](methyl)amine

  • MDL number MFCD09972215
  • Cas Number : 1017782-51-2
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94495

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart