KEY158583305 Key Organics / BIONET

{[5-(4-chlorophenyl)-1,3,4-oxadiazol-2-yl]methyl}(methyl)amine

  • MDL number MFCD09972216
  • Cas Number : 1017782-52-3
  • Molecular Weight : 223.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56 - 58
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94496

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart