KEY158583309 Key Organics / BIONET

2-[4-(2-hydroxyethyl)piperazin-1-yl]-1-(thiophen-2-yl)propan-1-one

  • MDL number MFCD02931014
  • Cas Number : 4506-87-0
  • Molecular Weight : 268.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94501

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart