KEY158583310 Key Organics / BIONET

{4'-nitro-[1,1'-biphenyl]-4-yl}methanol

  • MDL number MFCD09972221
  • Cas Number : 62037-99-4
  • Molecular Weight : 229.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94507

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0825_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart