KEY158583311 Key Organics / BIONET

2'-methanesulfinyl-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD09972222
  • Cas Number : 1171155-44-4
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94508

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0829_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart