KEY158583312 Key Organics / BIONET

1-(3-bromo-1-benzofuran-2-yl)ethan-1-one

  • MDL number MFCD09972223
  • Cas Number : 99661-02-6
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 89 - 91
Iupac Name 1-(3-bromo-1-benzofuran-2-yl)ethanone
Inchi Code 1S/C10H7BrO2/c1-6(12)10-9(11)7-4-2-3-5-8(7)13-10/h2-5H,1H3
Inchi Key QADCPIOQDOBMKS-UHFFFAOYSA-N

Description

Melting Point 89 - 91
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94509

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AC-0830_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart