KEY158583374 Key Organics / BIONET

2-[4-(4-methoxybenzenesulfonyl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD01211979
  • Cas Number : 321531-40-2
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 121 - 123
Iupac Name 2-{4-[(4-methoxyphenyl)sulfonyl]-1-piperazinyl}ethanol
Inchi Code 1S/C13H20N2O4S/c1-19-12-2-4-13(5-3-12)20(17,18)15-8-6-14(7-9-15)10-11-16/h2-5,16H,6-11H2,1H3
Inchi Key VEWVVGUOHKZQOI-UHFFFAOYSA-N

Description

Melting Point 121 - 123
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102125

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MB-0730_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart