KEY158583386 Key Organics / BIONET

2-methoxy-4-(morpholin-4-yl)aniline

  • MDL number MFCD09972210
  • Cas Number : 209960-91-8
  • Molecular Weight : 208.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78 - 80
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94486

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart