KEY158583392 Key Organics / BIONET

3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-2-carboxylic acid

  • MDL number MFCD01569306
  • Cas Number : 141783-63-3
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 95
Iupac Name 3-(tert-butoxycarbonyl)-1,3-thiazolidine-2-carboxylic acid
Inchi Code 1S/C9H15NO4S/c1-9(2,3)14-8(13)10-4-5-15-6(10)7(11)12/h6H,4-5H2,1-3H3,(H,11,12)
Inchi Key HYAXPNDMEODKHI-UHFFFAOYSA-N

Description

Melting Point 95
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102143

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MB-0821_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart