KEY158583400 Key Organics / BIONET

2-benzyl-1lambda6,2,5-thiadiazolidine-1,1-dione

  • MDL number MFCD04037043
  • Cas Number : 144432-72-4
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39 - 46
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101744

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LB-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart