KEY158583404 Key Organics / BIONET

4-(3-methoxy-4-nitrophenyl)morpholine

  • MDL number MFCD00023315
  • Cas Number : 6950-88-5
  • Molecular Weight : 238.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 168
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106669

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NB-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart