KEY158583412 Key Organics / BIONET

4-chloro-2-phenyl-6-(piperidin-1-yl)pyrimidine

  • MDL number MFCD00754197
  • Cas Number : 574708-35-3
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94490

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart