KEY158583413 Key Organics / BIONET

4-fluoro-1H-indazol-3-amine

  • MDL number MFCD00119934
  • Cas Number : 404827-78-7
  • Molecular Weight : 151.144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-5015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart