KEY159853848 Key Organics / BIONET

6-methyl-5-[(pyrrolidin-1-yl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD10758054
  • Cas Number : 1033463-16-9
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >300
Iupac Name 6-methyl-5-(1-pyrrolidinylmethyl)-2,4(1H,3H)-pyrimidinedione
Inchi Code 1S/C10H15N3O2/c1-7-8(6-13-4-2-3-5-13)9(14)12-10(15)11-7/h2-6H2,1H3,(H2,11,12,14,15)
Inchi Key WQEOCELMACWGTL-UHFFFAOYSA-N

Description

Melting Point >300
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94478

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AC-0031_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart