KEY159853849 Key Organics / BIONET

3-(4-chlorophenyl)-1H-pyrazol-4-amine

  • MDL number MFCD10758055
  • Cas Number : 91857-91-9
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136 - 138
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94483

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0221_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart