KEY159853856 Key Organics / BIONET

methyl 1-cyanocyclobutane-1-carboxylate

  • MDL number MFCD10758063
  • Cas Number : 58920-79-9
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name methyl 1-cyanocyclobutanecarboxylate
Inchi Code 1S/C7H9NO2/c1-10-6(9)7(5-8)3-2-4-7/h2-4H2,1H3
Inchi Key WCTZTNWBOZFXTE-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95006

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BC-0812_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart