KEY159853862 Key Organics / BIONET

4-bromo-2-fluoro-5-nitrobenzaldehyde

  • MDL number MFCD10758067
  • Cas Number : 679839-39-5
  • Molecular Weight : 248.007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47 - 49
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95625

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CC-0606_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart