KEY159853882 Key Organics / BIONET

5-bromo-3-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD10758088
  • Cas Number : 79456-34-1
  • Molecular Weight : 241.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 85 - 87
Iupac Name 5-bromo-3-(trifluoromethyl)-2-pyridinylamine
Inchi Code 1S/C6H4BrF3N2/c7-3-1-4(6(8,9)10)5(11)12-2-3/h1-2H,(H2,11,12)
Inchi Key QUYZBNHTYCLZLW-UHFFFAOYSA-N

Description

Melting Point 85 - 87
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96085

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DC-0818_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart