KEY159853913 Key Organics / BIONET

3-(2-methyl-1H-indol-3-yl)-3-oxopropanenitrile

  • MDL number MFCD02330570
  • Cas Number : 76397-72-3
  • Molecular Weight : 198.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94510

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0832_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart