KEY159853925 Key Organics / BIONET

2-methyl-3-nitropyridine

  • MDL number MFCD04114137
  • Cas Number : 18699-87-1
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 32 - 33
Iupac Name 2-methyl-3-nitropyridine
Inchi Code 1S/C6H6N2O2/c1-5-6(8(9)10)3-2-4-7-5/h2-4H,1H3
Inchi Key CCFGTKQIRWHYTB-UHFFFAOYSA-N

Description

Melting Point 32 - 33
Purity 95%
Physical Form Liquid,Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95624

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CC-0602_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart