KEY159853941 Key Organics / BIONET

methyl 3'-nitro-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD01846431
  • Cas Number : 89900-93-6
  • Molecular Weight : 257.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 146
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94497

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart