KEY159853944 Key Organics / BIONET

2-chloro-4-nitrobenzonitrile

  • MDL number MFCD00052203
  • Cas Number : 28163-00-0
  • Molecular Weight : 182.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-nitrobenzonitrile
Inchi Code 1S/C7H3ClN2O2/c8-7-3-6(10(11)12)2-1-5(7)4-9/h1-3H
Inchi Key ZIGQFRQZYVQQDR-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99827

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-188C_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart