KEY159853950 Key Organics / BIONET

5-(chloromethyl)-2-(trifluoromethyl)pyridine

  • MDL number MFCD01862650
  • Cas Number : 386715-33-9
  • Molecular Weight : 195.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Storage temp. RT
Physical Form Off-white to Dark Yellow Liquid,Low Melting Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101735

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LB-0736_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart