KEY180877463 Key Organics / BIONET

3-bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD11100204
  • Cas Number : 866546-09-0
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 240
Iupac Name 3-bromo-5-chloro-1H-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C7H4BrClN2/c8-6-3-11-7-5(6)1-4(9)2-10-7/h1-3H,(H,10,11)
Inchi Key MWDJROWZDUZXBP-UHFFFAOYSA-N

Description

Melting Point 240
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95655

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CC-0827_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart