KEY180877465 Key Organics / BIONET

[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methanol

  • MDL number MFCD11100207
  • Cas Number : 1033463-31-8
  • Molecular Weight : 211.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36 - 38
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96074

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DC-0730_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart