KEY180877480 Key Organics / BIONET

2-cyano-2,2-dimethylacetic acid

  • MDL number MFCD11100210
  • Cas Number : 22426-30-8
  • Molecular Weight : 113.116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153 - 155
Purity 95
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96093

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/DC-0839_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart