KEY180877487 Key Organics / BIONET

4-amino-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD08234883
  • Cas Number : 366452-98-4
  • Molecular Weight : 148.165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200 - 202
Purity 97
Storage temp. RT
Physical Form White to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96376

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/EC-0727_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart