KEY180877491 Key Organics / BIONET

4-nitro-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD09701290
  • Cas Number : 366452-97-3
  • Molecular Weight : 178.147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225 - 230
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96375

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EC-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart