KEY181739806 Key Organics / BIONET

2-amino-6-bromoquinolin-4-ol

  • MDL number MFCD08234781
  • Cas Number : 123420-09-7
  • Molecular Weight : 239.072

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94984

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BC-0204_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart