KEY181739808 Key Organics / BIONET

6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09258865
  • Cas Number : 1018828-69-7
  • Molecular Weight : 230.146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 239
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96373

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EC-0718_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart