KEY181739816 Key Organics / BIONET

5-bromo-2-ethyl-1,3-benzoxazole

  • MDL number MFCD09055180
  • Cas Number : 938458-80-1
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 64 - 66
Iupac Name 5-bromo-2-ethyl-1,3-benzoxazole
Inchi Code 1S/C9H8BrNO/c1-2-9-11-7-5-6(10)3-4-8(7)12-9/h3-5H,2H2,1H3
Inchi Key FEFKLDDJMPFJHS-UHFFFAOYSA-N

Description

Melting Point 64 - 66
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96845

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FC-0721_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart